Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008245.3 | NP_032271.1 | 2 | 533 | ||
GenBank mRNA | AK014111.1 | - | 2 | 532 | ||
AK033664.1 | - | 2 | 534 | |||
AK149553.1 | - | 2 | 534 | |||
BC057986.1 | - | 2 | 520 | 5124938 |