Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008248.2 | NP_032274.1 | 2 | 228 | ||
GenBank mRNA | AK002965.2 | - | 2 | 228 | ||
AK012433.1 | - | 2 | 234 | |||
BC070415.1 | - | 2 | 273 | 5714321 | ||
BC080296.1 | - | 2 | 212 | 6814185 | ||
U60001.1 | - | 2 | 198 |