Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122950.2 | NP_001116422.1 | 2 | 607 | ||
NM_008263.3 | NP_032289.2 | 2 | 1011 | |||
GenBank mRNA | AK002670.1 | - | 2 | 159 | ||
BC050839.1 | - | 2 | 983 | 6511608 | ||
L08757.1 | - | 2 | 1011 |