Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079869.1 | NP_001073338.1 | 4 | 1391 | ||
NM_010458.2 | NP_034588.2 | 2 | 1314 | |||
GenBank mRNA | S35628.1 | - | 2 | 1314 | ||
S35738.1 | - | 4 | 1392 | |||
X66177.1 | - | 2 | 1314 |