Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008270.2 | NP_032296.2 | 2 | 882 | ||
GenBank mRNA | AI116698.1 | - | 2 | 263 | ||
AK078860.1 | - | 1 | 281 | |||
BC100743.1 | - | 2 | 450 | 40045678 | ||
BC103573.1 | - | 2 | 450 | 40045679 | ||
M34857.1 | - | 2 | 301 | |||
S66855.1 | - | 2 | 703 |