Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013553.2 | NP_038581.2 | 2 | 1101 | ||
GenBank mRNA | AB221549.1 | - | 2 | 773 | ||
AK141658.1 | - | 4 | 1686 | |||
BC144778.1 | - | 2 | 1054 | 9053326 | ||
BC150664.1 | - | 2 | 1054 | 9087575 | ||
D11329.1 | - | 2 | 780 | |||
X69019.1 | - | 2 | 1100 |