Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 4 | 521 | ||
NM_001122897.2 | NP_001116369.1 | 5 | 758 | |||
NM_001285478.1 | NP_001272407.1 | 5 | 778 | |||
NM_001285479.1 | NP_001272408.1 | 5 | 719 | |||
NM_001285480.1 | NP_001272409.1 | 4 | 482 | |||
NM_001285481.1 | NP_001272410.1 | 5 | 680 | |||
NM_010470.3 | NP_034600.2 | 5 | 719 | |||
GenBank mRNA | AK004938.1 | - | 5 | 696 | ||
AK028181.1 | - | 5 | 697 | |||
AK043260.1 | - | 4 | 495 | |||
AK050234.1 | - | 5 | 698 | |||
AK075725.1 | - | 4 | 460 | |||
AK150784.1 | - | 5 | 764 | |||
AK161041.1 | - | 4 | 489 | |||
AK161084.1 | - | 5 | 613 | |||
AK166161.1 | - | 5 | 666 | |||
AK169740.1 | - | 5 | 625 | |||
AK212728.1 | - | 1 | 80 | |||
BC020024.1 | - | 5 | 669 | 3597151 | ||
BC107687.1 | - | 5 | 654 | |||
BU612149.1 | - | 4 | 522 | |||
X99642.1 | - | 3 | 362 |