Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021877.3 | NP_068677.2 | 4 | 1574 | ||
GenBank mRNA | AB221597.1 | - | 2 | 880 | ||
AY312855.1 | - | 3 | 1257 | |||
AY547390.1 | - | 3 | 1257 | |||
AY547391.1 | - | 3 | 1257 | |||
BC049182.1 | - | 4 | 1574 | 6468299 | ||
Z32675.1 | - | 3 | 1257 |