Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080999.2 | NP_001074468.1 | 10, 11 | 1860 | ||
NM_001081000.2 | NP_001074469.1 | 10, 11 | 1907 | |||
GenBank mRNA | AK034102.1 | - | 7, 8 | 2056 | ||
AK133844.1 | - | 10, 11 | 1735 | |||
AK134368.1 | - | 10, 11 | 1861 | |||
AK146391.1 | - | 10, 11 | 1800 | |||
AK150482.1 | - | 10, 11 | 1705 | |||
AK151311.1 | - | 10, 11 | 1705 | |||
AK151422.1 | - | 10, 11 | 1731 | |||
AK151725.1 | - | 10, 11 | 1705 | |||
AK151778.1 | - | 10, 11 | 1705 | |||
AK152055.1 | - | 10, 11 | 1705 | |||
AK152201.1 | - | 10, 11 | 1705 | |||
AK152363.1 | - | 10, 11 | 1705 | |||
AK152548.1 | - | 10, 11 | 1705 | |||
AK152903.1 | - | 10, 11 | 1705 | |||
AK153043.1 | - | 10, 11 | 1705 | |||
AK153190.1 | - | 10, 11 | 1731 | |||
BC027431.1 | - | 9, 10 | 1972 | 3156359 | ||
BC038486.1 | - | 10, 11 | 1733 | 3155739 | ||
X56044.1 | - | 9, 10 | 2304 |