Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008321.2 | NP_032347.1 | 2 | 383 | ||
GenBank mRNA | AK002820.1 | - | 2 | 381 | ||
AK031499.1 | - | 496 | ||||
BC145871.1 | - | 2 | 383 | 40131174 | ||
BC145873.1 | - | 2 | 383 | 40131176 | ||
M60523.1 | - | 2 | 383 |