Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111274.1 | NP_001104744.1 | 2 | 328 | ||
NM_001111275.1 | NP_001104745.1 | 2 | 410 | |||
NM_001111276.1 | NP_001104746.1 | 2 | 328 | |||
NM_010512.4 | NP_034642.2 | 2 | 410 | |||
NM_184052.3 | NP_908941.1 | 2 | 328 | |||
GenBank mRNA | AF440694.1 | - | 1 | 38 | ||
AK038119.1 | - | 2 | 1411 | |||
AK081019.1 | - | 2 | 399 | |||
AK155435.1 | - | 3 | 882 | |||
AK165471.1 | - | 2 | 300 | |||
AY878192.1 | - | 2 | 208 | |||
AY878193.1 | - | 2 | 208 | |||
BC012409.1 | - | 1 | 65 | 4194295 | ||
CA494190.1 | - | 451 | ||||
CT010364.1 | - | 1 | 38 | |||
X04480.1 | - | 2 | 190 | |||
X04482.1 | - | 2 | 110 |