Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010558.1 | NP_034688.1 | 3 | 329 | ||
GenBank mRNA | BC125366.1 | - | 3 | 314 | 40129881 | |
BC132066.1 | - | 3 | 314 | 40130343 | ||
X04601.1 | - | 3 | 329 | |||
X06270.1 | - | 3 | 329 |