Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253822.1 | NP_001240751.1 | 2 | 1657 | ||
NM_008393.3 | NP_032419.2 | 2 | 1657 | |||
GenBank mRNA | AK143023.1 | - | 2 | 1559 | ||
BC067195.1 | - | 2 | 1656 | |||
BC085500.1 | - | 2 | 1377 | 30539191 | ||
Y15001.1 | - | 2 | 1243 |