Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159417.1 | NP_001152889.1 | 3 | 510 | ||
NM_001159418.1 | NP_001152890.1 | 2 | 308 | |||
NM_008394.3 | NP_032420.1 | 2 | 424 | |||
GenBank mRNA | AK145911.1 | - | 1 | 176 | ||
AK149742.1 | - | 1 | 175 | |||
BC005435.1 | - | 3 | 285 | 3601255 | ||
BC012968.1 | - | 2 | 319 | 3257714 | ||
CK788642.1 | - | 2 | 251 | |||
U51992.1 | - | 2 | 228 |