Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008427.4 | NP_032453.3 | 2 | 869 | ||
GenBank mRNA | AB024304.1 | - | 1 | 754 | ||
BC120593.1 | - | 2 | 846 | 40129516 | ||
BC137594.1 | - | 2 | 846 | 8860613 | ||
S71382.1 | - | 2 | 870 | |||
U11075.1 | - | 2 | 839 |