Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008429.2 | NP_032455.2 | 3 | 912 | ||
GenBank mRNA | AF130860.1 | - | 3 | 875 | ||
AK015907.1 | - | 3 | 958 | |||
AK033970.1 | - | 3 | 912 | |||
AK046084.1 | - | 3 | 951 | |||
BC065161.1 | - | 2 | 713 | 5718431 | ||
U11860.1 | - | 3 | 961 |