Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010616.2 | NP_034746.1 | 19 | 2041 | ||
GenBank mRNA | AB755406.1 | - | 19 | 2057 | ||
AK018598.1 | - | 19 | 2043 | |||
BC022225.1 | - | 14 | 3116 | 4222522 |