Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010617.2 | NP_034747.2 | 26 | 3384 | ||
GenBank mRNA | AK029721.1 | - | 8 | 1020 | ||
AK077053.1 | - | 26 | 3408 |