Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008444.4 | NP_032470.3 | 5, 6 | 1899 | ||
GenBank mRNA | AK083700.1 | - | 5, 6 | 1887 | ||
AK122258.1 | - | 5, 6 | 1860 | |||
AK147467.1 | - | 5, 6 | 1893 | |||
AK148200.1 | - | 5, 6 | 1880 | |||
AK161946.1 | - | 3, 4 | 339 | |||
BC139388.1 | - | 4, 5 | 1818 | 8862410 | ||
BC139389.1 | - | 4, 5 | 1818 | 8862411 | ||
D26077.1 | - | 4, 5 | 1812 |