Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291222.1 | NP_001278151.1 | 15 | 3339 | ||
NM_010626.3 | NP_034756.2 | 15 | 3336 | |||
GenBank mRNA | BC034093.1 | - | 5 | 911 | ||
BC141045.1 | - | 15 | 3337 | 9055657 | ||
BC145222.1 | - | 15 | 3340 | 9053773 |