Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272097.1 | NP_001259026.1 | 3 | 363 | ||
NM_001272098.1 | NP_001259027.1 | 2 | 190 | |||
NM_010634.3 | NP_034764.1 | 3 | 363 | |||
GenBank mRNA | AK008782.1 | - | 3 | 357 | ||
AK011551.1 | - | 3 | 359 | |||
AK166506.1 | - | 3 | 356 | |||
AK167389.1 | - | 3 | 358 | |||
AK216545.1 | - | 2 | 172 | |||
BC002008.1 | - | 3 | 343 | 3490535 | ||
BC053042.1 | - | 1 | 2923 | |||
BC100543.1 | - | 3 | 338 | 30136900 | ||
W54445.1 | - | 2 | 143 | |||
X70100.1 | - | 3 | 342 |