Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008457.2 | NP_032483.1 | 5 | 671 | ||
GenBank mRNA | BC031798.1 | - | 5 | 676 | 4167600 | |
X03994.1 | - | 5 | 670 |