Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008458.2 | NP_032484.1 | 3 | 785 | ||
GenBank mRNA | BC116724.1 | - | 3 | 783 | 40106394 |