Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159374.2 | NP_001152846.2 | 4 | 762 | ||
GenBank mRNA | AK009595.1 | - | 2 | 89 | ||
BC117553.1 | - | 4 | 652 | 40104816 | ||
BC118064.1 | - | 4 | 652 | 40104817 | ||
X75649.1 | - | 4 | 652 |
Gene Symbol: Gm11571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028798.1 | - | 3 | 1459 |