Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ktn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293635.1 | NP_001280564.1 | 33 | 3284 | ||
NM_001293636.1 | NP_001280565.1 | 34 | 3404 | |||
NM_008477.2 | NP_032503.2 | 33 | 3242 | |||
GenBank mRNA | AJ517365.1 | - | 10 | 649 | ||
AJ517366.1 | - | 10 | 650 | |||
AJ517367.1 | - | 10 | 650 | |||
AJ517368.1 | - | 10 | 649 | |||
AJ517369.1 | - | 10 | 650 | |||
AJ517370.2 | - | 10 | 650 | |||
AJ517371.1 | - | 11 | 718 | |||
AJ517372.1 | - | 11 | 719 | |||
AJ517373.1 | - | 11 | 737 | |||
AJ517374.1 | - | 11 | 737 | |||
AJ517375.1 | - | 12 | 806 | |||
AJ517376.1 | - | 10 | 650 | |||
AJ517377.1 | - | 11 | 737 | |||
AJ517378.1 | - | 10 | 649 | |||
AJ517379.1 | - | 10 | 650 | |||
AJ517380.1 | - | 11 | 737 | |||
AJ517381.1 | - | 11 | 737 | |||
AJ517382.1 | - | 11 | 702 | |||
AJ517383.1 | - | 12 | 806 | |||
AJ517384.1 | - | 10 | 650 | |||
AK172869.1 | - | 9 | 1053 | |||
BC066871.1 | - | 33 | 3239 | |||
BC075697.1 | - | 34 | 3323 | |||
CD363975.1 | - | 2 | 95 | |||
L43326.1 | - | 33 | 3230 |