Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181564.2 | NP_853543.1 | 16 | 2537 | ||
GenBank mRNA | AB240192.1 | - | 15 | 2720 | ||
AY191584.1 | - | 16 | 2537 | |||
AY227349.1 | - | 16 | 2403 | |||
BC109009.2 | - | 16 | 2537 | |||
BC150475.2 | - | 16 | 2537 |