Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010690.1 | NP_034820.1 | 8 | 3750 | ||
GenBank mRNA | AF104414.1 | - | 5 | 2875 | ||
BC158092.1 | - | 7 | 3596 | 9088177 | ||
BC158123.1 | - | 8 | 3745 | 9088196 |