Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010691.5 | NP_034821.2 | 1 | 122 | ||
GenBank mRNA | AB108497.1 | - | 1 | 122 | ||
BC119177.1 | - | 1 | 211 | 8733926 | ||
BC120586.1 | - | 1 | 211 | 40129509 | ||
X90829.1 | - | 1 | 611 |