Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136069.2 | NP_001129541.2 | 7 | 1110 | ||
NM_010699.2 | NP_034829.1 | 7 | 917 | |||
GenBank mRNA | AK089929.1 | - | 7 | 881 | ||
AK146121.1 | - | 7 | 878 | |||
AK146980.1 | - | 3 | 292 | |||
AK149919.1 | - | 7 | 878 | |||
AK150156.1 | - | 7 | 881 | |||
AK167789.1 | - | 7 | 879 | |||
AK167944.1 | - | 7 | 879 | |||
AK168346.1 | - | 7 | 878 | |||
AK168920.1 | - | 7 | 883 | |||
AK170386.1 | - | 7 | 882 | |||
AK170708.1 | - | 7 | 879 | |||
AK171373.1 | - | 6 | 762 | |||
AK177468.1 | - | 3 | 275 | |||
AK188760.1 | - | 3 | 276 | |||
AK198750.1 | - | 2 | 159 | |||
AK200149.1 | - | 3 | 226 | |||
AK203954.1 | - | 2 | 159 | |||
AK216074.1 | - | 2 | 159 | |||
AK217915.1 | - | 2 | 159 | |||
AK220005.1 | - | 2 | 160 | |||
BC005509.1 | - | 6 | 729 | |||
BC066858.1 | - | 6 | 535 | |||
BC094019.1 | - | 7 | 834 | 4195220 | ||
BC094428.1 | - | 7 | 871 | 30700835 | ||
U13687.1 | - | 7 | 977 |