Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013581.3 | NP_038609.3 | 7 | 1592 | ||
GenBank mRNA | AF109377.2 | - | 7 | 1575 | ||
AK031510.1 | - | 6 | 1048 | |||
AK034636.2 | - | 7 | 1597 | |||
AK152464.1 | - | 7 | 1583 | |||
AK158507.1 | - | 7 | 1954 | |||
AK159497.1 | - | 7 | 1594 | |||
AK162971.1 | - | 7 | 1593 | |||
AK171574.1 | - | 7 | 1586 | |||
AK220387.1 | - | 7 | 1682 | |||
BC063056.1 | - | 7 | 1574 | 5720460 |