Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lect1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010701.2 | NP_034831.2 | 6, 7 | 959 | ||
GenBank mRNA | AK013975.1 | - | 6, 7 | 885 | ||
BC045152.1 | - | 6, 7 | 858 | 6439864 | ||
U43509.1 | - | 6, 7 | 960 |