Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010712.2 | NP_034842.2 | 6 | 1158 | ||
GenBank mRNA | AB221565.1 | - | 5 | 939 | ||
AF135415.1 | - | 5 | 943 | |||
BC049834.1 | - | 6 | 1158 | 6493203 | ||
S71659.1 | - | 3 | 528 |