Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034030.2 | NP_001029202.1 | 7, 8 | 850 | ||
NM_010718.4 | NP_034848.1 | 11, 12 | 1489 | |||
NM_173053.1 | NP_774958.1 | 10, 11 | 1620 | |||
GenBank mRNA | AB005131.1 | - | 11, 12 | 1342 | ||
AB005132.1 | - | 10, 11 | 1604 | |||
AB008117.1 | - | 11, 12 | 1425 | |||
AB012092.1 | - | 7, 8 | 850 | |||
AB012291.1 | - | 7, 8 | 850 | |||
AK034900.1 | - | 2, 3 | 151 | |||
AK146099.1 | - | 10, 11 | 1577 | |||
BC007129.1 | - | 11, 12 | 1432 | 3496836 | ||
BC086629.1 | - | 10, 11 | 1550 |