Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013585.2 | NP_038613.1 | 5 | 504 | ||
GenBank mRNA | AK008429.1 | - | 5 | 495 | ||
AK152743.1 | - | 5 | 549 | |||
AK171670.1 | - | 5 | 514 | |||
BC032210.1 | - | 3 | 2519 | |||
BC116364.2 | - | 5 | 504 | 40100021 | ||
CT010227.1 | - | 5 | 483 | |||
D44454.1 | - | 5 | 504 | |||
D44456.1 | - | 5 | 504 | |||
D44457.1 | - | 5 | 504 | |||
D44458.1 | - | 5 | 504 | |||
D44461.1 | - | 5 | 504 | |||
KJ650226.1 | - | 5 | 483 | |||
S58203.1 | - | 5 | 504 | |||
S59862.1 | - | 5 | 504 | |||
U22447.1 | - | 5 | 483 | |||
U22448.1 | - | 5 | 483 | |||
U22919.1 | - | 5 | 483 | |||
U22920.1 | - | 5 | 483 |