Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008514.4 | NP_032540.2 | 15 | 3448 | ||
GenBank mRNA | AF074265.1 | - | 15 | 3317 | ||
AK140796.1 | - | 3 | 843 | |||
AK162203.1 | - | 2 | 252 | |||
BC060704.1 | - | 15 | 3448 | 6409058 |