Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010734.2 | NP_034864.2 | 3, 4 | 196 | ||
GenBank mRNA | AI182980.1 | - | 3, 4 | 196 | ||
AI323110.1 | - | 2, 3 | 101 | |||
AI482475.1 | - | 1, 2 | 219 | |||
AK146409.1 | - | 2, 3 | 54 | |||
BC086881.1 | - | 3, 4 | 190 | 6747449 | ||
M18187.1 | - | 3, 4 | 189 | |||
U72643.1 | - | 3, 4 | 185 | |||
U72644.1 | - | 3, 4 | 185 |