Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008518.2 | NP_032544.1 | 1, 2 | 176 | ||
GenBank mRNA | BC064062.1 | - | 1, 2 | 160 | ||
BC114919.1 | - | 1, 2 | 156 | |||
U16985.1 | - | 1, 2 | 161 |