Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130450.2 | NP_569717.1 | 4, 5 | 513 | ||
GenBank mRNA | AB072039.1 | - | 4, 5 | 460 | ||
AF480860.1 | - | 4, 5 | 432 | |||
AK029029.1 | - | 4, 5 | 515 | |||
AK165847.1 | - | 3, 4 | 537 | |||
AK167373.1 | - | 4, 5 | 479 | |||
AY053453.1 | - | 4, 5 | 494 | |||
BC040097.1 | - | 4, 5 | 455 | |||
BC051041.1 | - | 4, 5 | 510 | 4012676 | ||
BC098492.1 | - | 3, 4 | 510 | 30091954 | ||
BC100576.1 | - | 4, 5 | 455 | 30097278 |