Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028621.3 | NP_082897.2 | 1 | 209 | ||
GenBank mRNA | AF345297.1 | - | 1 | 203 | ||
AK003736.2 | - | 1 | 240 | |||
BC145974.1 | - | 1 | 209 | |||
BC145976.1 | - | 1 | 209 | |||
BC147776.1 | - | 1 | 303 | 9087497 | ||
BC147786.1 | - | 1 | 303 | 9087505 |
Gene Symbol: Krtap6-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010673.2 | NP_034803.2 | 1 | 218 | ||
GenBank mRNA | AA727286.1 | - | 1 | 215 | ||
BC116356.1 | - | 1 | 217 | |||
BC116357.1 | - | 1 | 217 | |||
BC128485.1 | - | 1 | 217 | |||
BC128486.1 | - | 1 | 217 | |||
D89902.1 | - | 2 | 230 |