Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133215.1 | NP_573478.1 | 9, 10 | 947 | ||
GenBank mRNA | AF262986.1 | - | 9, 10 | 947 | ||
AK146641.1 | - | 9, 10 | 964 | |||
BC058091.1 | - | 9, 10 | 945 | 6822365 | ||
BC058364.1 | - | 9, 10 | 945 | 6820754 |