Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133218.2 | NP_573481.1 | 5 | 1404 | ||
GenBank mRNA | AF292939.1 | - | 5 | 1404 | ||
BC052936.1 | - | 2 | 138 |