Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081346.1 | NP_001074815.1 | 2, 3 | 414 | ||
GenBank mRNA | AK030986.1 | - | 2, 3 | 298 | ||
AK045134.1 | - | 315 |