Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008535.2 | NP_032561.2 | 4 | 1282 | ||
GenBank mRNA | AK076114.1 | - | 4 | 936 | ||
BC005736.1 | - | 4 | 902 | 3598911 | ||
CO044699.1 | - | 1 | 533 | |||
CT010167.1 | - | 3 | 727 | |||
X57687.1 | - | 4 | 1192 |