Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 5, 6 | 735 | ||
GenBank mRNA | AK004656.1 | - | 4, 5 | 2378 | ||
AK133943.1 | - | 5, 6 | 741 | |||
AK145913.1 | - | 5, 6 | 747 | |||
AK149177.1 | - | 5, 6 | 765 | |||
AK151607.1 | - | 5, 6 | 796 | |||
AK153140.1 | - | 5, 6 | 747 | |||
AK153302.1 | - | 5, 6 | 816 | |||
AK153474.1 | - | 5, 6 | 756 | |||
AK196390.1 | - | 1, 2 | 123 | |||
AK197278.1 | - | 1, 2 | 123 | |||
BC027210.1 | - | 5, 6 | 690 | |||
BC046956.1 | - | 5, 6 | 735 | |||
BC080811.1 | - | 5, 6 | 735 | 6438831 | ||
M63286.1 | - | 5, 6 | 643 | |||
X56831.1 | - | 5, 6 | 675 | |||
X64068.1 | - | 5, 6 | 738 |
Gene Symbol: M6pr-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X64069.1 | - | 1 | 1101 |