Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010751.3 | NP_034881.2 | 4, 5 | 474 | ||
GenBank mRNA | AK151014.1 | - | 4, 5 | 436 | ||
AK151114.1 | - | 4, 5 | 435 | |||
AK151305.1 | - | 4, 5 | 473 | |||
AK151882.1 | - | 4, 5 | 435 | |||
AK153303.1 | - | 4, 5 | 435 | |||
BC034860.1 | - | 4, 5 | 411 | 1448816 | ||
CT010233.1 | - | 4, 5 | 323 | |||
L38926.1 | - | 4, 5 | 359 | |||
U20614.1 | - | 4, 5 | 428 | |||
X83106.1 | - | 4, 5 | 372 |