Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134188.3 | NP_598949.3 | 1 | 108 | ||
GenBank mRNA | AK141651.1 | - | 1 | 98 | ||
AK172617.1 | - | 1 | 104 | |||
BC064469.1 | - | 1 | 79 | 6508650 |