Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Madcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013591.2 | NP_038619.2 | 5 | 1226 | ||
GenBank mRNA | AK137028.1 | - | 5 | 1227 | ||
AK156096.1 | - | 4 | 772 | |||
BC024372.1 | - | 4 | 787 | 4986802 | ||
BC115910.1 | - | 4 | 788 | 40105157 | ||
BC118056.1 | - | 4 | 788 | 40105156 | ||
L21203.1 | - | 5 | 1223 | |||
S80258.1 | - | 4 | 761 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK077345.1 | - | 1877 | Chr. - - [-] on Build |