Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134245.1 | NP_599006.1 | 1 | 878 | ||
GenBank mRNA | BC145650.1 | - | 1 | 956 | 40130953 |