Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199349.1 | NP_001186278.1 | 4 | 510 | ||
NM_138309.3 | NP_612182.1 | 3 | 441 | |||
GenBank mRNA | AK031264.1 | - | 4 | 267 | ||
AK054364.1 | - | 4 | 310 | |||
AK080225.1 | - | 4 | 266 | |||
AK166275.1 | - | 4 | 261 | |||
AY078163.1 | - | 3 | 237 | |||
BC031736.1 | - | 3 | 216 | 4013657 | ||
BY727540.1 | - | 4 | 511 | |||
EF516990.1 | - | 4 | 268 |