Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282737.1 | NP_001269666.1 | 4 | 396 | ||
GenBank mRNA | AF007862.1 | - | 4 | 383 | ||
AF035939.1 | - | 4 | 343 | |||
AK011512.1 | - | 4 | 335 | |||
BC018176.1 | - | 4 | 334 | 3587774 |